Order Entry
Export
ContactUsLinkComponent
3-Bromothiophene
3-Bromothiophene
Catalog # MOLEM82813398
Supplier:  Molekula
CAS Number:  
3-Bromothiophene
Catalog # MOLEM82813398
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :163.04 g/mol
Boiling Pt:158 °C (1013 hPa)
Melting Pt:–10 °C
Density:1.735 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:212-821-3
UN:2810
ADR:6.1,II
CAS Number:872-31-1
Formula:C₄H₃BrS
MDL Number:MFCD00005464