Order Entry
Export
ContactUsLinkComponent
3,4-Dibromothiophene
Irritant
3,4-Dibromothiophene
Catalog # MOLE28121389-5G
Supplier:  Molekula
CAS Number:  
Irritant
3,4-Dibromothiophene
Catalog # MOLE28121389-5G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :241.93 g/mol
Boiling Pt:221 °C (1013 hPa)
Boiling Pt:221 °C (1013 hPa)
Melting Pt:4.5 °C
Melting Pt:4,5 °C
Density:2,13 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:221-546-8
 :GHS07
CAS Number:3141-26-2
Formula:C₄H₂Br₂S
MDL Number:MFCD00005465