Order Entry
Export
ContactUsLinkComponent
 
Irritant
2,5-Dibromothiophene
Catalog # MOLEM82396898
Supplier:  Molekula
CAS Number:  
Irritant
2,5-Dibromothiophene
Catalog # MOLEM82396898
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :241.93 g/mol
Boiling Pt:211 °C (1013 hPa)
Melting Pt:–6 °C
Density:2.1531 g/cm³ (13.8 °C)
EINECS:221-547-3
UN:2810
ADR:6.1,II
 :GHS07
CAS Number:3141-27-3
Formula:C₄H₂Br₂S
MDL Number:MFCD00005420