Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,5-Dibromothiophene
2,5-Dibromothiophene 96%
2,5-Dibromothiophene 96%
:
Thermo Fisher Scientific
2,5-Dibromothiophene 96%
:
Thermo Fisher Scientific
:
3141-27-3
:
:
C₄H₂Br₂S
:
MFCD00005420
:
241.93 g/mol
:
210…212 °C
:
–6 °C
:
2.150
:
99 °C (210 °F)
:
Ambient
:
221-547-3
:
2810
:
6.1,II
:
GHS06
:
GHS08
:
GHS07
msds_ALFAA13406
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years
Thermo Scientific Organic Photovoltaics