2,4-Dibromothiophene
Catalog # MOLE12518574-1G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:241.93 g/mol
Boiling Pt:215 °C (1013 hPa)
Melting Pt:–30 °C
Density:2.14 g/cm³ (20 °C)
UN:2810
ADR:6.1,III
CAS Number:3140-92-9
Formula:C₄H₂Br₂S
MDL Number:MFCD00043889