2,4-Dibromothiophene
Supplier: Thermo Scientific
Certificates
About this item
CAS Number:3140-92-9
:
Formula:C₄H₂Br₂S
MDL Number:MFCD00043889
:241.93 g/mol
Boiling Pt:215 °C (1013 hPa)
Melting Pt:–30 °C
Density:2.14 g/cm³ (20 °C)
UN:2810
ADR:6.1,III