

2,4-Dibromothiophene ≥90%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3140-92-9
:
Formula:C₄H₂Br₂S
MDL Number:MFCD00043889
:241.93 g/mol
Boiling Pt:215 °C
Melting Pt:–30 °C
Density:2.160
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
UN:2810
ADR:6.1,III
:GHS06
:GHS07