2,3-Dibromothiophene
Supplier: Thermo Scientific
Jump to:
About this item
- CAS Number:3140-93-0
- Formula:C₄H₂Br₂S
- MDL Number:MFCD00005418
- Molecular Weight:241.93 g/mol
- Boiling Pt:130 °C (1013 hPa)
- Melting Pt:–17.5 °C
- Density:2.2 g/cm³ (20 °C)
- EINECS:221-542-6
- UN:1993
- ADR:3,III
- :Density: 2.2 g/cm³ (20 °C)
2 Options Available Below
Product Details & Documents
- CAS Number:3140-93-0
- Formula:C₄H₂Br₂S
- MDL Number:MFCD00005418
- Molecular Weight:241.93 g/mol
- Boiling Pt:130 °C (1013 hPa)
- Melting Pt:–17.5 °C
- Density:2.2 g/cm³ (20 °C)
- EINECS:221-542-6
- UN:1993
- ADR:3,III
- :Density: 2.2 g/cm³ (20 °C)
Product Family Options
Product Information
- Pack typePkAvailabilityPrice



