
2,6-Diphenylpyridine ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3558-69-8
:
Formula:C₁₇H₁₃N
MDL Number:MFCD00006284
:231.3 g/mol
Boiling Pt:210 °C (3 mmHg)
Melting Pt:74…81 °C
Storage Temperature:Ambient
EINECS:222-620-2
:GHS07