

2,4,6-Collidine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:108-75-8
:
Formula:C₈H₁₁N
MDL Number:MFCD00006338
Merck Index:13,09789
Boiling Pt:171 °C (1013 hPa)
Melting Pt:–43 °C
Density:0.9191 g/cm³ (20 °C)
Flash Pt:55 °C
UN:1992
ADR:3,III
:121.18 g/mol
EINECS:203-613-3
:GHS02
:GHS07
Storage Temperature:Ambient