

2,4,6-Collidine 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:108-75-8
:
Formula:C₈H₁₁N
MDL Number:MFCD00006338
Merck Index:13,09789
Boiling Pt:171…172 °C (760 mmHg)
Melting Pt:–43 °C
Density:0.91 g/cm³
Flash Pt:57 °C
UN:1992
ADR:3,III
:121.18 g/mol
EINECS:203-613-3
:GHS02
:GHS06
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Clear colorless to yellow Liquid (May darken on storage)
- Infrared spectrum:Conforms
- Refractive index:1.4970 to 1.4990 (20°C, 589 nm)
- GC:≥98.5 %
- Sensitivity:(to chlorine) Passes test