2,6-Lutidine
Glass bottle for solids, 1 kg
Catalog # MOLE84306154
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: 2,6-Dimethylpyridine
- CAS Number:108-48-5
- Formula:C₇H₉N
- MDL Number:MFCD00006345
- Molecular Weight:107.16 g/mol
- Merck Index:13,05636
- Boiling Pt:143.7 °C (1013 hPa)
- Melting Pt:–6 °C
- Density:0.9252 g/cm³ (20 °C)
- Flash Pt:33 °C
- Storage Temperature:Ambient
- EINECS:203-587-3
- UN:1993
- ADR:3,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg



