
2,6-Di-tert-butylpyridine
Catalog # MOLE15293583-5G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:191.32 g/mol
Merck Index:12,03086
Boiling Pt:209 °C (1013 hPa)
Density:0.852 g/cm³ (25 °C)
EINECS:209-557-6
:GHS07
Storage Temperature:Refrigerator
CAS Number:585-48-8
Formula:C₁₃H₂₁N
MDL Number:MFCD00006306