2,3-Lutidine
Glass bottle for solids, 100 g
Catalog # MOLE22252417-100G
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: 2,3-Dimethylpyridine
- CAS Number:583-61-9
- Formula:C₇H₉N
- MDL Number:MFCD00009605
- Molecular Weight:107.16 g/mol
- Boiling Pt:161 °C (1013 hPa)
- Melting Pt:–15 °C
- Density:0.945 g/cm³ (20 °C)
- EINECS:209-514-1
- UN:2929
- ADR:6.1,II
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



