2,3-Lutidine 98+%
Glass bottle, 25 mL
: 2,3-Dimethylpyridine
- :583-61-9
- :C₇H₉N
- :MFCD00009605
- :107.16 g/mol
- :161 °C (760 mmHg)
- :–17 °C
- :0.95 g/cm³
- :49 °C
- :Ambient
- :209-514-1
- :1993
- :3,III
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5062 to 1.5082 (20°C, 589 nm)
- Water:≤0.5 %
- GC:Impurities profile
- Pack type:Glass bottle
- Pk:25 mL



