
2,6-Diaminopyridine 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:141-86-6
:
Formula:C₅H₇N₃
MDL Number:MFCD00006329
:109.13 g/mol
Boiling Pt:285 °C (760 mmHg)
Melting Pt:115…123 °C
Flash Pt:155 °C
Storage Temperature:Ambient
EINECS:205-507-2
UN:2811
ADR:6.1,II
:GHS06
Specification Test Results
- Appearance:Beige to grey to dark brown Crystalline powder or flakes (May darken on storage)
- Infrared spectrum:Conforms
- Melting point:115 to 123 °C
- Water:≤1 %
- GC:≥97.5 %