Order Entry
Export
Orders LinkContactUsLinkComponent
2,3-Diaminopyridine

2,3-Diaminopyridine

Glass bottle for solids, 5 g
Catalog # MOLEM59418972
Irritant
Supplier:   Molekula
2,3-Diaminopyridine
Catalog # MOLEM59418972
Supplier:  Molekula
CAS Number:  452-58-4
2,3-Diaminopyridine

Glass bottle for solids, 5 g

CAS Number:  452-58-4
Price
Inquire For Price
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  Pyridine-2,3-diyldiamine
  • CAS Number:452-58-4
  • Formula:C₅H₇N₃
  • MDL Number:MFCD00006319
  • Molecular Weight:109.13 g/mol
  • Boiling Pt:148…150 °C (5 mmHg)
  • Melting Pt:110…112 °C
  • Storage Temperature:Ambient
  • EINECS:207-200-9
  • UN:2671
  • ADR:6.1,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications