Order Entry
Export
Orders LinkContactUsLinkComponent
2,3-Diaminopyridine 98%

2,3-Diaminopyridine 98%

Toxic
Irritant
2,3-Diaminopyridine 98%
Certificates
2,3-Diaminopyridine 98%
2,3-Diaminopyridine 98%

3 Options Available Below

Product Details & Documents

Purity: 98% CAS No.: 452-58-4 Molecular Formula: C5H7N3 Formula Weight: 109.13 Flash Point: 205°(401°F) Melting Point: 110-114° Boiling Point: 195°/55mm Beilstein Reference: 109869 UN No.: 2671 MDL No.: MFCD00006319

Synonyms:  Pyridine-2,3-diyldiamine
  • CAS Number:452-58-4
  • Formula:C₅H₇N₃
  • MDL Number:MFCD00006319
  • Molecular Weight:109.13 g/mol
  • Boiling Pt:195 °C (55 mmHg)
  • Melting Pt:110…114 °C
  • Flash Pt:205 °C (401 °F)
  • Storage Temperature:Ambient
  • EINECS:207-200-9
  • UN:2671
  • ADR:6.1,II
Product Family Options

Product Information