2,6-Difluoropyridine 99%
- :1513-65-1
- :C₅H₃F₂N
- :MFCD00006227
- :115.08 g/mol
- :124.5 °C (743 mmHg)
- :1.26 g/cm³
- :33 °C
- :Ambient
- :216-152-8
- :1993
- :3,III
- :1513-65-1
- :C₅H₃F₂N
- :MFCD00006227
- :115.08 g/mol
- :124.5 °C (743 mmHg)
- :1.26 g/cm³
- :33 °C
- :Ambient
- :216-152-8
- :1993
- :3,III
- Appearance:Clear colorless to yellow to pink to red-purple Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4360 to 1.4390 (20°C, 589 nm)
- GC:≥98.5 %
Product Family Options
- Pack typePk



