


3,5-Dichloro-2,4,6-trifluoropyridine ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1737-93-5
:
Formula:C₅Cl₂F₃N
MDL Number:MFCD00006226
:201.96 g/mol
Boiling Pt:159…160 °C
Melting Pt:24…26 °C
Density:1.648
EINECS:217-088-3
UN:2811
ADR:6.1,II
:GHS06
:GHS05
:GHS07
Storage Temperature:Ambient