2,6-Dichloropyridine
Glass bottle for solids, 100 g
Catalog # MOLE34574238-100G
Supplier: Molekula
Jump to:
Product Details & Documents
- CAS Number:2402-78-0
- Formula:C₅H₃Cl₂N
- MDL Number:MFCD00006244
- Molecular Weight:147.99 g/mol
- Boiling Pt:211 °C (1013 hPa)
- Melting Pt:83…86 °C
- Storage Temperature:Ambient
- EINECS:219-282-3
- UN:2811
- ADR:6.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



