2,6-Dichloropyridine 98%
Supplier: Thermo Fisher Scientific
About this item
- CAS Number:2402-78-0
- Formula:C₅H₃Cl₂N
- MDL Number:MFCD00006244
- Molecular Weight:147.99 g/mol
- Boiling Pt:211 °C (1013 hPa)
- Melting Pt:84…89 °C
- Flash Pt:110 °C
- Storage Temperature:Ambient
- EINECS:219-282-3
- ADR:6.1,III
2 Options Available Below
Product Details & Documents
- CAS Number:2402-78-0
- Formula:C₅H₃Cl₂N
- MDL Number:MFCD00006244
- Molecular Weight:147.99 g/mol
- Boiling Pt:211 °C (1013 hPa)
- Melting Pt:84…89 °C
- Flash Pt:110 °C
- Storage Temperature:Ambient
- EINECS:219-282-3
- ADR:6.1,III
Specifications for Product Family
Specification Test Results
- Appearance:White to pink to light grey Crystalline powder
- Infrared spectrum:Conforms
- Melting point:84 to 89 °C
- GC:≥97.5 %
- Iodate (IO3):≤3 ppm
- Phosphate (PO4):≤0.001 %
- Sulfate (SO4):≤0.005 %
- Barium (Ba):≤0.002 %
- Calcium (Ca):≤0.002 %
- Iron (Fe):≤5 ppm
- Magnesium (Mg):≤0.001 %
- Potassium (K):≤0.01 %
- Heavy metals (as Pb):≤5 ppm
- Insoluble matter:≤0.01 %



