2,4-Dichloropyridine 98%
Supplier: Thermo Fisher Scientific
About this item
- CAS Number:26452-80-2
- Formula:C₅H₃Cl₂N
- MDL Number:MFCD00955618
- Molecular Weight:147.99 g/mol
- Melting Pt:–1 °C
- Density:1.37 g/cm³
- Flash Pt:89 °C
- Storage Temperature:Ambient
- EINECS:247-717-7
- ADR:6.1,III
2 Options Available Below
Product Details & Documents
- CAS Number:26452-80-2
- Formula:C₅H₃Cl₂N
- MDL Number:MFCD00955618
- Molecular Weight:147.99 g/mol
- Melting Pt:–1 °C
- Density:1.37 g/cm³
- Flash Pt:89 °C
- Storage Temperature:Ambient
- EINECS:247-717-7
- ADR:6.1,III
Specifications for Product Family
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid (May darken on storage)
- Infrared spectrum:Conforms
- Refractive index:1.5500 to 1.5540 (20°C, 589 nm)
- GC:≥97.5 %



