
3,5-Dibromopyridine
Catalog # MOLEM88427567
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
Boiling Pt:222 °C (1013 hPa)
:236.89 g/mol
EINECS:210-916-4
:GHS07
Storage Temperature:Ambient
CAS Number:625-92-3
Formula:C₅H₃Br₂N
MDL Number:MFCD00014634
Boiling Pt:222 °C (1013 hPa)