
3,5-Dibromopyridine 98+%
:625-92-3
:
:C₅H₃Br₂N
:MFCD00014634
:222 °C (1013 hPa)
:110…114 °C
:>115 °C
:Ambient
:210-916-4
:2811
:6.1,III
:236.89 g/mol
:GHS07
- Appearance:White to light beige Powder or crystals or needles
- Infrared spectrum:Conforms
- Melting point:110 to 114 °C
- GC:≥98.0 %