Order Entry
Export
Orders LinkContactUsLinkComponent
 

2-Bromopyridine

Glass bottle for solids, 100 g
Catalog # MOLE28504127-100G
Supplier:   Molekula
2-Bromopyridine
Catalog # MOLE28504127-100G
Supplier:  Molekula
CAS Number:  109-04-6
2-Bromopyridine

Glass bottle for solids, 100 g

CAS Number:  109-04-6
Price
180,00 €
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:109-04-6
  • Formula:C₅H₄BrN
  • MDL Number:MFCD00006219
  • Molecular Weight:158 g/mol
  • Boiling Pt:192 °C (1013 hPa)
  • Melting Pt:–40.1 °C
  • Density:1.62 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:203-641-6
  • UN:2810
  • ADR:6.1,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications