2-Bromopyridine 99%
Supplier: Thermo Fisher Scientific
About this item
- CAS Number:109-04-6
- Formula:C₅H₄BrN
- MDL Number:MFCD00006219
- Molecular Weight:158 g/mol
- Boiling Pt:192…194 °C (761 mmHg)
- Melting Pt:-40,1 °C
- Density:1.65 g/cm³
- Flash Pt:54 °C
- Storage Temperature:Ambient
- EINECS:203-641-6
- UN:2929
- ADR:6.1,II
3 Options Available Below
Product Details & Documents
- CAS Number:109-04-6
- Formula:C₅H₄BrN
- MDL Number:MFCD00006219
- Molecular Weight:158 g/mol
- Boiling Pt:192…194 °C (761 mmHg)
- Melting Pt:-40,1 °C
- Density:1.65 g/cm³
- Flash Pt:54 °C
- Storage Temperature:Ambient
- EINECS:203-641-6
- UN:2929
- ADR:6.1,II
Specifications for Product Family
Specification Test Results
- Appearance:Clear colorless to yellow to light brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5700 to 1.5730 (20°C, 589 nm)
- GC:≥98.5 %
- Color scale:≤20 APHA



