2,6-Dibromopyridine
Glass bottle for solids, 25 g
Catalog # MOLE19976185-25G
Supplier: Molekula
Jump to:
Product Details & Documents
- CAS Number:626-05-1
- Formula:C₅H₃Br₂N
- MDL Number:MFCD00006223
- Molecular Weight:236.89 g/mol
- Boiling Pt:255 °C (1013 hPa)
- Melting Pt:116…119 °C
- Storage Temperature:Ambient
- EINECS:210-926-9
- UN:2811
- ADR:6.1,II
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



