Tetrahydrofuran
Supplier: Roth Carl
Certificates
About this item
Synonyms: Butylene oxide, THF, Diethylene oxide
CAS Number:109-99-9
:
Formula:C₄H₈O
MDL Number:MFCD00005356
Boiling Pt:66
Melting Pt:–108.5 °C
Density:0.888 g/cm³ (20 °C)
Molecular Weight:72.11 g/mol
Flash Pt:–21.5 °C
UN:2056
ADR:3 II
Merck Index:12,09351
Storage Temperature:Ambient
EINECS:203-726-8
REACH:05-2114615188-48
:GHS02
:GHS08
:GHS07
Specifications
- Environmentally Preferable: