Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,2'-Binaphthyl
2,2'-Binaphthyl
2,2'-Binaphthyl
2,2'-Binaphthyl
Supplier:
CHEMPUR
Certificates
Expand image
2,2'-Binaphthyl
Supplier:
CHEMPUR
About this item
CAS Number:
612-78-2
:
Formula:
C₂₀H₁₄
MDL Number:
MFCD00021584
:
254.33 g/mol
Boiling Pt:
452 °C (1013 hPa)
Melting Pt:
188.3 °C
Density:
1.14 g/cm³ (20 °C)
EINECS:
210-320-4