
2,3-Dimethylnaphthalene
Catalog # EHERC20772000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 mg
- Seq:2275
Specifications
About this item
:
:156.23 g/mol
Boiling Pt:269 °C (1013 hPa)
Melting Pt:103…104 °C
Density:1,003 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:209-463-5
:GHS07
CAS Number:581-40-8
Formula:C₁₂H₁₂
MDL Number:MFCD00004119