1,7-Dimethylnaphthalene
Catalog # EHERC20767000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 mg
- Seq:2315
Specifications
About this item
:
:156.23 g/mol
Boiling Pt:263 °C (1013 hPa)
Melting Pt:–13 °C
Density:1.0115 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:209-382-5
CAS Number:575-37-1
Formula:C₁₂H₁₂
MDL Number:MFCD00060884