Order Entry
Export
ContactUsLinkComponent
1,6-Dimethylnaphthalene
1,6-Dimethylnaphthalene
Catalog # EHERC20765000
Supplier:  EHRENSTORFER
CAS Number:  
1,6-Dimethylnaphthalene
Catalog # EHERC20765000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
Boiling Pt:265 °C (1013 hPa)
Melting Pt:–16.9 °C
Density:1,009 g/cm³ (20 °C)
 :156.23 g/mol
Melting Pt:–16,9 °C
Storage Temperature:Ambient
EINECS:209-385-1
CAS Number:575-43-9
Formula:C₁₂H₁₂
MDL Number:MFCD00004039