Order Entry
Export
ContactUsLinkComponent
1,3-Dimethylnaphthalene
1,3-Dimethylnaphthalene
Catalog # EHERC20755000
Supplier:  EHRENSTORFER
CAS Number:  
1,3-Dimethylnaphthalene
Catalog # EHERC20755000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 mg
  • Seq:
    2370

Specifications

About this item

 :
Boiling Pt:263 °C (1013 hPa)
Melting Pt:–4 °C
Density:1.0146 g/cm³ (20 °C)
 :156.23 g/mol
Storage Temperature:Ambient
EINECS:209-384-6
CAS Number:575-41-7
Formula:C₁₂H₁₂
MDL Number:MFCD00004036