Order Entry
Export
ContactUsLinkComponent
1,2-Dimethylnaphthalene
1,2-Dimethylnaphthalene
Catalog # EHERC20750000
Supplier:  EHRENSTORFER
CAS Number:  
1,2-Dimethylnaphthalene
Catalog # EHERC20750000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 mg
  • Seq:
    2370

Specifications

About this item

 :
 :156.23 g/mol
Boiling Pt:268 °C (1013 hPa)
Boiling Pt:268 °C (1013 hPa)
Density:1.0219 g/cm³ (16.3 °C)
Storage Temperature:Ambient
EINECS:209-364-7
CAS Number:573-98-8
Formula:C₁₂H₁₂
MDL Number:MFCD00004035