1,2-Dimethylnaphthalene ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:573-98-8
:
Formula:C₁₂H₁₂
MDL Number:MFCD00004035
:156.23 g/mol
Boiling Pt:266…267 °C
Density:1.013
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:209-364-7