1-Naphthol ≥99%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 99% CAS No.: 90-15-3 Molecular Formula: C10H8O Formula Weight: 144.17 Flash Point: 125°(257°F) Odour: schwach, phenolartig Boiling Point: 278-280° Solubility: Soluble in alcohol, benzene, chloroform, ether Beilstein Reference: 1817321 Merck Reference: 14,6383 UN No.: 2811 MDL No.: MFCD00003930
Synonyms: 1-Hydroxynaphthalene, α-Naphthol, α-Hydroxynaphthalene
- CAS Number:90-15-3
- Formula:C₁₀H₈O
- MDL Number:MFCD00003930
- Molecular Weight:144.17 g/mol
- Merck Index:14,6383
- Boiling Pt:278…280 °C
- Melting Pt:95…97 °C
- Density:1.28
- Flash Pt:125 °C (257 °F)
- Storage Temperature:Ambient
- EINECS:201-969-4
- UN:2811
- ADR:6.1,III
3 Options Available Below
Product Details & Documents
Purity: 99% CAS No.: 90-15-3 Molecular Formula: C10H8O Formula Weight: 144.17 Flash Point: 125°(257°F) Odour: schwach, phenolartig Boiling Point: 278-280° Solubility: Soluble in alcohol, benzene, chloroform, ether Beilstein Reference: 1817321 Merck Reference: 14,6383 UN No.: 2811 MDL No.: MFCD00003930
Synonyms: 1-Hydroxynaphthalene, α-Naphthol, α-Hydroxynaphthalene
- CAS Number:90-15-3
- Formula:C₁₀H₈O
- MDL Number:MFCD00003930
- Molecular Weight:144.17 g/mol
- Merck Index:14,6383
- Boiling Pt:278…280 °C
- Melting Pt:95…97 °C
- Density:1.28
- Flash Pt:125 °C (257 °F)
- Storage Temperature:Ambient
- EINECS:201-969-4
- UN:2811
- ADR:6.1,III



