Order Entry
Export
Orders LinkContactUsLinkComponent
1-Naphthol 99+%

1-Naphthol 99+%

Irritant
Corrosive
1-Naphthol 99+%
Certificates
1-Naphthol 99+%

3 Options Available Below

Product Details & Documents
Synonyms:  1-Hydroxynaphthalene, α-Naphthol, α-Hydroxynaphthalene
  • CAS Number:90-15-3
  • Formula:C₁₀H₈O
  • MDL Number:MFCD00003930
  • Molecular Weight:144.17 g/mol
  • Merck Index:13,06409
  • Boiling Pt:278…280 °C (760 mmHg)
  • Melting Pt:95…97 °C
  • Density:1,224 g/cm³ (20 °C)
  • Flash Pt:125 °C
  • Storage Temperature:Ambient
  • EINECS:201-969-4
  • UN:2811
  • ADR:6.1,III

Documents

Certificates
Search by Lot Number
Specifications for Product Family

Specification Test Results

  • Appearance:
    Beige to grey to brown Crystalline powder or crystals or flakes
  • Infrared spectrum:
    Conforms
  • Melting point:
    95 to 97 °C
  • GC:
    ≥99.0 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 27,30 €
    27,30 €
    1 * 100 g
       27,30 €

  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 71,90 €
    71,90 €
    1 * 500 g
       71,90 €

  • Plastic bottle
    2,5 kg
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 266,00 €
    266,00 €
    1 * 2,5 kg
       266,00 €