Order Entry
Export
ContactUsLinkComponent
1,8-Dinitronaphthalene
1,8-Dinitronaphthalene
Catalog # EHERC20974800
Supplier:  EHRENSTORFER
CAS Number:  
1,8-Dinitronaphthalene
Catalog # EHERC20974800
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
 :218.17 g/mol
Boiling Pt:390 °C (1013 hPa)
Boiling Pt:390 °C (1013 hPa)
Melting Pt:173 °C
EINECS:210-016-1
CAS Number:602-38-0
Formula:C₁₀H₆N₂O₄
MDL Number:MFCD00003918