Order Entry
Export
Orders LinkContactUsLinkComponent
1-Nitronaphthalene

1-Nitronaphthalene

Glass bottle for solids, 500 g
Catalog # MOLEM29344002
Supplier:   Molekula
1-Nitronaphthalene
Catalog # MOLEM29344002
Supplier:  Molekula
CAS Number:  86-57-7
1-Nitronaphthalene

Glass bottle for solids, 500 g

CAS Number:  86-57-7
Price
Inquire For Price
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  α-Nitronaphthalene
  • CAS Number:86-57-7
  • Formula:C₁₀H₇NO₂
  • MDL Number:MFCD00003913
  • Molecular Weight:173.17 g/mol
  • Merck Index:12,06711
  • Boiling Pt:304 °C (1013 hPa)
  • Melting Pt:59…61 °C
  • Density:1.331 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:201-684-5
  • UN:2538
  • ADR:4.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications