Order Entry
Export
Orders LinkContactUsLinkComponent
Zinc oxide

Zinc oxide

Environment
Supplier:   Roth Carl
Zinc oxide
Supplier:  Roth Carl
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  Zinci oxidum, Zinc (II) oxide
  • CAS Number:1314-13-2
  • Formula:ZnO
  • MDL Number:MFCD00011300
  • Molecular Weight:81.39 g/mol
  • Merck Index:13,10200
  • Melting Pt:1975 °C
  • Density:5.61 g/cm³ (25 °C)
  • Storage Temperature:Ambient
  • EINECS:215-222-5
  • UN:3077
  • ADR:9,III
  • REACH:01-2119463881-32
Product Family Options

Product Information

  • Pk
    Availability
    Price
  • 500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 85,10 €
    85,10 €
    1 * 500 g
       85,10 €

  • 1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 134,00 €
    134,00 €
    1 * 1 kg
       134,00 €