Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Zinc oxide
Zinc oxide heavy 98%
Zinc oxide heavy 98%
:
Thermo Fisher Scientific
Zinc oxide heavy 98%
:
Thermo Fisher Scientific
:
1314-13-2
:
:
ZnO
:
MFCD00011300
:
81.39 g/mol
:
14,10147
:
1975 °C
:
5.606
:
Ambient
:
215-222-5
:
3077
:
9,III
:
01-2119463881-32
:
GHS09
msds_ALFAA16188
Thermo Scientific Acroseal 25 Years