Order Entry
Export
Orders LinkContactUsLinkComponent
 

Zinc oxide 99.5+%

Environment
Zinc oxide 99.5+%
Certificates
Zinc oxide 99.5+%

2 Options Available Below

Product Details & Documents
Synonyms:  Zinci oxidum, Zinc (II) oxide
  • CAS Number:1314-13-2
  • Formula:ZnO
  • MDL Number:MFCD00011300
  • Molecular Weight:81.39 g/mol
  • Merck Index:13,10200
  • Melting Pt:1975 °C
  • Density:5.6 g/cm³
  • Storage Temperature:Ambient
  • EINECS:215-222-5
  • UN:3077
  • ADR:9,III
  • REACH:01-2119463881-32

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    White to faintly yellowish-white Powder
  • Titration Complexometric:
    ≥99.50 %
  • Additional info:
    Carbonates and substances insoluble in acids:
  • Particle size:
    ≤0.01 % Sieve residue on 45 micron sieve
  • Residue after ignition:
    ≤1 % (1 g, 500°C)
  • Specific surface area:
    3.5 to 9 m²/g
  • Alkalinity:
    ≤0.3 ml/g (ml 0.1 N HCl/g)
  • Arsenic (As):
    ≤5 ppm
  • Cadmium (Cd):
    ≤10 ppm
  • Iron (Fe):
    ≤200 ppm
  • Lead (Pb):
    ≤50 ppm
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Plastic bottle
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 58,80 €
    58,80 €
    1 * 1 kg
       58,80 €

  • Plastic drum
    5 kg
    Log in for availability
    Log in for availability
    1 * 5 kg
    1 * 5 kg - 201,00 €
    201,00 €
    1 * 5 kg
       201,00 €