Order Entry
Export
Orders LinkContactUsLinkComponent
 

Silver nitrate

Oxidizing
Corrosive
Environment
Supplier:   Roth Carl
Silver nitrate
Supplier:  Roth Carl
Certificates
Silver nitrate

4 Options Available Below

Product Details & Documents
Synonyms:  Lunar caustic, Silver mononitrate, Nitric acid silver salt
  • CAS Number:7761-88-8
  • Formula:AgNO₃
  • MDL Number:MFCD00003414
  • Molecular Weight:169.87 g/mol
  • Merck Index:12,08661
  • Boiling Pt:444 °C (1013 hPa)
  • Melting Pt:210 °C
  • Density:4.35 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-853-9
  • UN:1493
  • ADR:5.1,II
  • REACH:01-2119513705-43
Product Family Options

Product Information

  • Pk
    Availability
    Price
  • 10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 43,50 €
    43,50 €
    1 * 10 g
       43,50 €

  • 50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g - 155,00 €
    155,00 €
    1 * 50 g
       155,00 €

  • 250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g - 649,00 €
    649,00 €
    1 * 250 g
       649,00 €

  • 1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 2 340,00 €
    2 340,00 €
    1 * 1 kg
       2 340,00 €