Order Entry
Export
Orders LinkContactUsLinkComponent
Silver nitrate 99+% ACS

Silver nitrate 99+% ACS

Oxidizing
Corrosive
Environment
Silver nitrate 99+% ACS
Certificates
Silver nitrate 99+% ACS

5 Options Available Below

Product Details & Documents
Synonyms:  Lunar caustic, Silver mononitrate, Nitric acid silver salt
  • CAS Number:7761-88-8
  • Formula:AgNO₃
  • MDL Number:MFCD00003414
  • Molecular Weight:169.87 g/mol
  • Merck Index:12,08661
  • Boiling Pt:444 °C (760 mmHg)
  • Melting Pt:212 °C
  • Density:4,35 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-853-9
  • UN:1493
  • ADR:5.1,II
  • REACH:01-2119513705-43

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    White Crystalline powder or crystals
  • Free acid:
    Passes test
  • Titration:
    ≥99.0 %
  • Clarity of solution:
    Passes test
  • Chloride (Cl):
    ≤5 ppm
  • Sulfate (SO4):
    ≤20 ppm
  • Copper (Cu):
    ≤2 ppm
  • Iron (Fe):
    ≤2 ppm
  • Lead (Pb):
    ≤10 ppm
  • Subst not pptd by HCl:
    ≤0.01 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g - 244,00 €
    244,00 €
    1 * 50 g
       244,00 €

  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 392,00 €
    392,00 €
    1 * 100 g
       392,00 €

  • Glass bottle for solids
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g - 918,00 €
    918,00 €
    1 * 250 g
       918,00 €

  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 1 550,00 €
    1 550,00 €
    1 * 500 g
       1 550,00 €

  • Glass bottle
    2,5 kg
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 6 000,00 €
    6 000,00 €
    1 * 2,5 kg
       6 000,00 €