5,6-Dimethoxy-1-indanone
Catalog # MOLE14744935-5G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:192.21 g/mol
Boiling Pt:302 °C (1013 hPa)
Melting Pt:120 °C
Storage Temperature:Ambient
EINECS:218-287-8
CAS Number:2107-69-9
Formula:C₁₁H₁₂O₃
MDL Number:MFCD00003790