
Diphosphorus pentoxide, Sigma-Aldrich®
Catalog # SIAL79609-500G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:141.94 g/mol
Merck Index:13,07441
Melting Pt:562 °C
Density:2.253 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-236-1
UN:1807
ADR:8,II
:GHS05
CAS Number:1314-56-3
Formula:P₂O₅
MDL Number:MFCD00011440