Order Entry
Export
ContactUsLinkComponent
Diphosphorus pentoxide
Corrosive
Diphosphorus pentoxide
Catalog # ROTH9076.2
Supplier:  Roth Carl
CAS Number:  
Corrosive
Diphosphorus pentoxide
Catalog # ROTH9076.2
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications

About this item

 :
 :141.94 g/mol
Merck Index:13,07441
Melting Pt:562 °C
Density:2.253 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-236-1
UN:1807
ADR:8,II
 :GHS05
CAS Number:1314-56-3
Formula:P₂O₅
MDL Number:MFCD00011440