Orders LinkContactUsLinkComponent
Accucore™ kits
Accucore™ kits
Accucore™ kits
Certificates
Accucore™ kits
6 Options Available Below
Product Details & Documents
Accucore™ kits validate the reproducibility of Accucore™ HPLC columns, or verify which Accucore™ HPLC column offers the best separation.
Accucore™ validation kit: Validate the reproducibility of Accucore™ HPLC columns.
Accucore™ narrow selectivity kit: Verify which Accucore™ HPLC column offers the best separation over a narrow selectivity range.
Accucore™ wide selectivity kit: Verify which Accucore™ HPLC column offers the best separation over a wide selectivity range.
Accucore™ polar selectivity kit: Verify which Accucore™ HPLC column offers the best separation for polar analytes.
Delivery information: The Accucore™ validation kit contains three Accucore™ C18 columns. The Accucore™ narrow selectivity kit contains one each of Accucore™ C18, RP-MS and aQ columns. The Accucore™ wide selectivity kit contains one each of Accucore™ C18, Phenyl-Hexyl and PFP columns. The Accucore™ polar selectivity kit contains one each of Accucore™ aQ, PFP and HILIC columns.
Product Family Options
Accucore™ validation kit
Product Information
  • Particle Size
    Ø int.
    Length
    Availability
    Price
  • 554-1960
    Validation kit
    Validation kit
    2,6 µm
    2,10 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 3 items
    1 * 3 items - 2 000,00 €
    2 000,00 €
    1 * 3 items
       2 000,00 €
  • 554-1961
    Validation kit
    Validation kit
    2,6 µm
    2,10 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 3 items
    1 * 3 items - 2 250,00 €
    2 250,00 €
    1 * 3 items
       2 250,00 €
  • 554-1962
    Validation kit
    Validation kit
    2,6 µm
    2,10 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 3 items
    1 * 3 items - 2 580,00 €
    2 580,00 €
    1 * 3 items
       2 580,00 €
Accucore™ narrow selectivity kit
Product Information
  • Particle Size
    Ø int.
    Length
    Availability
    Price
  • 554-1963
    Narrow selectivity kit
    Narrow selectivity kit
    2,6 µm
    2,10 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 3 items
    1 * 3 items - 2 060,00 €
    2 060,00 €
    1 * 3 items
       2 060,00 €
Accucore™ polar selectivity kit
Product Information
  • Particle Size
    Ø int.
    Length
    Availability
    Price
  • 554-1970
    Polar selectivity kit
    Polar selectivity kit
    2,6 µm
    2,10 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 3 items
    1 * 3 items - 2 250,00 €
    2 250,00 €
    1 * 3 items
       2 250,00 €
  • 554-1971
    Polar selectivity kit
    Polar selectivity kit
    2,6 µm
    2,10 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 3 items
    1 * 3 items - 2 780,00 €
    2 780,00 €
    1 * 3 items
       2 780,00 €