Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Cysteaminium chloride
Cysteaminium chloride, Sigma-Aldrich®
Cysteaminium chloride, Sigma-Aldrich®
Cysteaminium chloride, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
Cysteaminium chloride, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
156-57-0
:
Formula:
C₂H₇NS·HCl
MDL Number:
MFCD00012904
:
113.61 g/mol
:
M-Clarity™ quality level = MQ200
EINECS:
205-858-1